missing translation for 'onlineSavingsMsg'
Learn More

Lymphotoxin beta Receptor, Biotin, clone: eBio3C8 (3C8), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5671-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209839

  • £68.82 / 25µg


EU Stock 1
Add to basket

Description

Description

Description: The antibody reacts with mouse lymphotoxin-beta receptor (LTBR). Northern blot analysis of tissues from adult mice showed that expression levels of LTBR mRNA were strong in lung, liver, and kidney, moderate in heart and testes, but weak in brain, thymus, spleen, and lymph nodes. The tumor necrosis factor receptor-related protein is the human receptor for the heterotrimer of lymphotoxin-alpha and lymphotoxin-beta. This LT-alpha/LT-beta heterotrimer (LT-alpha1beta2) is assumed to take part in immunologic reactions by cell-cell contact, but does not bind to either TNFR1 (CD120alpha) or TNFR2 (CD120beta). LTBR is expressed by day 7 embryos, so it has been speculated that the LT-alpha/LT-beta receptor system may also have some function in early embryogenesis.Applications Reported: This eBio3C8 (3C8) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio3C8 (3C8) antibody has been tested by flow cytometric analysis of NIH/3T3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended to block Fc gamma-receptors with anti-mouse CD16/32 (cat. 14-0161), prior to staining to prevent non-specific binding of eBio3C8. Furthermore, it is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Lymphotoxin beta Receptor
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P50284
CD18, D12S370, LT-BETA-R, TNF-R-III, TNFCR, TNFR-RP, TNFR2-RP, TNFRSF3
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBio3C8 (3C8)
Biotin
Ltbr
Conjugated
Ltbr
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
17000
Videos

For Research Use Only.