Learn More
LY6G5B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561705
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CCQYDYCNSW SSPQLQSSLP EPHDRPLALP LSDSQIQWFY QALNLSLPLP NFHAGTEPDG LDPMVTLSLN LGLSFAELRR MYLFLNSSGL LVLPQ Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 61%.
LY6G5B Polyclonal antibody specifically detects LY6G5B in Human samples. It is validated for Immunohistochemistry.Specifications
LY6G5B | |
Polyclonal | |
Unconjugated | |
LY6G5B | |
C6orf19; G5B; LY6G5B; Lymphocyte antigen 6 complex locus protein G5b; lymphocyte antigen 6 complex, locus G5B; lymphocyte antigen 6 family member G5B; lymphocyte antigen-6 G5B splicing isoform 288; lymphocyte antigen-6 G5B splicing isoform 452; lymphocyte antigen-6 G5B splicing isoform 690; lymphocyte antigen-6 G5B splicing isoform 837 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58496 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NDX9 | |
LY6G5B | |
Recombinant protein corresponding to Human LY6G5B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only