missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-49I Monoclonal Antibody (YLI-90), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15414
Code : NEW
Additional Details : Weight : 0.00280kg
Description
Ly-49I Monoclonal antibody specifically detects Ly-49I in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-49I | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
0 | |
Klra9 | |
CHO-K1 cells transfected with the B6 allele of the Ly-49I gene, K1ra9 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
YLI-90 | |
PE | |
Klra9 | |
5E6; Killer cell lectin-like receptor 3; killer cell lectin-like receptor subfamily A, member 9; Klra3; Klra9; Ly-49; Ly49C; Ly-49c; Ly49I; Ly49I receptor; Ly-49I receptor; LY49I1; Lymphocyte antigen 49c; Nk2.1; T-cell surface glycoprotein Ly-49C | |
Mouse | |
purified | |
RUO | |
16640 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |