missing translation for 'onlineSavingsMsg'
Learn More

Ly-6K, PE, clone: B33fc8k, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5929-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15248919

  • £143.47 / 100µg


Estimated Shipment date
from Supplier 24-07-2018
Add to basket

Description

Description

Description: This B33fc8k monoclonal antibody reacts with mouse Ly-6k, a member of the Ly6/uPAR superfamily. Ly-6k is a GPI linked protein predominantly found on the cell surface. One study suggests expression may also be non-membranous. Expression is found on B cells and reported to be on plasma cells as well as cells in the testis. Using the B33fc8k monoclonal antibody, expression is on a B lymphocyte cell subset in the spleen that correlates to transitional cells (CD23-IgM-). In humans Ly-6k has been found associated with carcinomas of the head and testis.Applications Reported: This B33fc8k antibody has been reported for use in flow cytometric analysis.Applications Tested: This B33fc8k antibody has been tested by flow cytometric analyis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ly-6K
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9CWP4
CO16, CT97, cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
B33fc8k
PE
Ly6k
Conjugated
Ly6k
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
76486
Videos

For Research Use Only.