missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat, PE-Cy7, Clone: 1A8, enQuireBio™
Rat Monoclonal Antibody
£89.95 - £265.00
Specifications
Antigen | Ly-6G |
---|---|
Clone | 1A8 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15972218
|
EnquireBio
QAB56-PE7-25UG |
25 μg |
£89.95
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962218
|
EnquireBio
QAB56-PE7-100UG |
100 μg |
£265.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Ly-6G | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
546644 | |
IgG2a κ | |
Protein A/G Purified |
1A8 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
XP_006521677 | |
Ly6g | |
Primary | |
Store at 2-4°C. |