missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G Rat anti-Mouse, PerCP-eFluor™ 710, Clone: 1A8-Ly6g, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-9668-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15569426

  • £200.00 / 100µg

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. Ly6G (Lymphocyte antigen 6 complex locus G6D) is a component of the myeloid differentiation antigen Gr-1, together with Ly6C. The Ly6G protein has a molecular weight of approximately 21-25 kDa and is expressed by myeloid derived cells in the bone marrow. Ly6G is a good marker for detection of peripheral neutrophils, monocytes and granulocytes. Expression of Gr-1 in bone marrow correlates with granulocyte differentiation and maturation. The physiological role of Ly6G remains unclear but studies have shown its involvement in anti-tumor response, and studies with neutrophils suggest that Ly6G ligation may have an inhibitory effect on local immune responses.
Specifications

Specifications

Ly-6G
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P35461
Gr-1, Gr1, Ly-6G
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
1A8-Ly6g
PerCP-eFluor™ 710
Ly6g
Liquid
Ly6g
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
546644
Videos
Documents

For Research Use Only.