Learn More
Ly-6G Monoclonal Antibody (1A8-Ly6g), PE-eFluor™ 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 61-9668-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal antibody 1A8-Ly6g reacts with mouse Ly-6G, which is also known as Gr-1. Ly-6G is a 25-kDa GPI protein expressed exclusively by neutrophils. Studies suggest an important role in neutrophil infiltration, recruitment and migration. Ly-6G associates with B2 integrins CD11a and CD11b and can attenuate B2 integrin expression and function. The ligand for Ly-6G is currently unknown. Unlike the RB6-8C5 antibody, 1A8-Ly6g is reported to only bind to Ly-6G found on neutrophils. Moreover, 1A8-Ly6g has been used to deplete neutrophils and reported to have antagonist activity. Applications Reported: This 1A8-Ly6g antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1A8-Ly6g antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome.
Ly-6G Monoclonal antibody specifically detects Ly-6G in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-6G | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P35461 | |
Ly6g | |
Affinity chromatography | |
RUO | |
546644 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
1A8-Ly6g | |
PE-eFluor 610 | |
Ly6g | |
Gr1; Gr-1; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6G | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.