missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G (Gr-1), FITC, clone: RB6-8C5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5931-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228299

  • £49.20 / 100µg


EU Stock 10
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets.Applications Reported: The RB6-8C5 antibody has been reported for use in flow cytometric analysis.Applications Tested: The RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ly-6G (Gr-1)
RB6-8C5
FITC
Conjugated
IgG2b
RUO
Primary
Murine
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos