Ly-6G (Gr-1) Rat anti-Mouse, Clone: RB6-8C5, eBioscience™

Rat Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 14-5931-81

50UG:ANTI-MOUSE LY-6G (GR-1) PURIFIED 50 UG

UNSPSC: 12352200

Code: Z2

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only.

Product Code. 15297597

Quantity Price
1 £ 29.97 / 50µg
Estimated Shipment date
from Supplier 29-03-2017
Add to basket


Description and Specification

Specification

Antigen Ly-6G (Gr-1)
Format Purified
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Host Species Rat
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2b
Quantity 50μg
Regulatory Status RUO
Species Reactivity Mouse
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone RB6-8C5
Concentration 0.5mg/mL
Conjugate Unlabeled

The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.

eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets.