missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G (Gr-1), Alexa Fluor 532, clone: RB6-8C5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 58-5931-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15332180

  • £74.36 / 25µg


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Description

Description

Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets.Applications Reported: This RB6-8C5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Alexa Fluor® 532 is excited with the Green laser (532 nm) and emits at 561 nm. This cannot be used with the Yellow-Green laser (561 nm). We recommend using a 560/14 band pass filter. Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 532 nm; Emission: 561 nm; Laser: Green Laser
Specifications

Specifications

Ly-6G (Gr-1)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P35461
Gr-1, Gr1, Ly-6G
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
RB6-8C5
Alexa Fluor 532
Ly6g
Conjugated
Ly6g
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
546644
Videos

For Research Use Only.