missing translation for 'onlineSavingsMsg'
Learn More

Ly-6D, PerCP-eFluor™ 710, clone: 49-H4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-5974-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15539326

  • £189.40 / 100µg


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This 49-H4 monoclonal antibody reacts with mouse Ly-6D, which is also known as ThB. This member of the Ly6 family of GPI-anchored cell surface glycoproteins is expressed on thymocytes and splenic B cells. Moreover, common lymphoid progenitor cells that express Ly-6D have been shown to commit to the B cell lineage. Expression of Ly-6D has also been associated with Rag-1 expression along with a reduced ability to differentiate into T or NK cells. Finally, reports indicate that the level of Ly-6D expression varies in different mouse strains.Applications Reported: This 49-H4 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 49-H4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ly-6D
49-H4
PerCP-eFluor 710
Conjugated
IgG2c
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Videos