Learn More
Ly-6C Monoclonal Antibody (HK1.4), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£109.00 - £247.00
Specifications
Antigen | Ly-6C |
---|---|
Clone | HK1.4 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15321200
|
Affymetrix eBioscience
45-5932-80 |
25 μg |
£109.00
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15331200
|
Affymetrix eBioscience
45-5932-82 |
100 μg |
£247.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody HK1.4 recognizes mouse Ly-6C, a GPI-linked protein of the Ly6 family. Ly-6C is found on monocytes/macrophages, endothelial cells and granulocytes as well as a subset of lymphocytes. Some variation of expression is found on different mouse strains in regards to expression on CD4 and CD8 lymphocytes. These correlate to 2 alleles both of which are recognized by HK1.4: Ly6c.1 found on C57Bl/6 and SJL cells which results in staining of both CD4 and CD8 cells while Ly6-C.2 found on BALB/c and 3H/He results in staining of CD8, but not CD4 cells. In vitro addition with HK1.4 antibody can increase proliferation and stimulate cytokine release. Applications Reported: This HK1.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HK1.4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Ly-6C Monoclonal antibody specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-6C | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AA682074; AA959465; Ly6c; Ly-6C; Ly6c1; Ly-6C1; Ly6c2; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6C1; Lymphocyte antigen Ly-6C | |
Ly6c1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Ly6c1 |
HK1.4 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
Rat | |
Mouse | |
P0CW02 | |
17067 | |
IgG2c κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.