Learn More
Ly-6A/E (Sca-1) Monoclonal Antibody (D7), PE-eFluor™ 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 61-5981-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The D7 monoclonal antibody reacts with mouse Sca-1, an 18 kDa member of the Ly-6 family of GPI-linked surface proteins. D7 reacts with both Ly-6E.1 and Ly-6A.2 molecules expressed by mouse hematopoietic stem cells, myeloid population and peripheral T and B cells. Non-activated splenocytes from Ly-6.2 strains including C57BL, SJL, 129, AKR, and others demonstrate higher frequency of D7-positive cells compared to the Ly-6.1 strains such as BALB/c, C3H, NZB and others. Sca-1 is expressed at high levels upon activation regardless of the Ly-6 haplotype. The Ly-6 family is involved in regulation and function of T cell activation. Sca-1 is a major phenotypic marker for mouse hematopoietic progenitor/stem cell subset. Applications Reported: This D7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This D7 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm.
Ly-6A/E (Sca-1) Monoclonal antibody specifically detects Ly-6A/E (Sca-1) in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-6A/E (Sca-1) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P05533, Q64253 | |
Ly6a, Ly6e | |
Affinity chromatography | |
RUO | |
110454, 17069 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
D7 | |
PE-eFluor 610 | |
Ly6a | |
9804; locus A/E; Locus AE; Ly6; Ly67; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly6A/E; Ly-6A/E; ly6a-e; Ly6e; Ly-6E; Ly-6E.1; lymphocyte antigen 6 complex; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6 complex, locus E; lymphocyte antigen 6A-2/6E-1; Lymphocyte antigen 6E; RIG-E; Sca1; Sca-1; Sca-2; stem cell antigen 1; Stem cell antigen 2; TAP; T-cell-activating protein; thymic shared antigen 1; Tsa1; TSA-1 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.