missing translation for 'onlineSavingsMsg'
Learn More

Ly-6A/E (Sca-1), PE, clone: D7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5981-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15269219

  • £124.59 / 100µg


EU Stock 8
Add to basket

Description

Description

Description: The D7 monoclonal antibody reacts with mouse Sca-1, an 18kDa member of the Ly-6 family of GPI-linked surface proteins. D7 reacts with both Ly-6E.1 and Ly-6A.2 molecules expressed by mouse hematopoietic stem cells, myeloid population and peripheral T and B cells. Non-activated splenocytes from Ly-6.2 strains including C57BL, SJL, 129, AKR, and others demonstrate higher frequency of D7-positive cells compared to the Ly-6.1 strains such as BALB/c, C3H, NZB and others. Sca-1 is expressed at high levels upon activation regardless of the Ly-6 haplotype. The Ly-6 family is involved in regulation and function of T cell activation. Sca-1 is a major phenotypic marker for mouse hematopoietic progenitor/stem cell subset.Applications Reported: This D7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This D7 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ly-6A/E (Sca-1)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q64253, P05533
lymphocyte antigen 6 complex, locus A/E
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
D7
PE
Ly6e
Conjugated
Ly6a, Ly6e
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
110454, 17069
Videos

For Research Use Only.