missing translation for 'onlineSavingsMsg'
Learn More

Ly-6A/E (Sca-1) Monoclonal Antibody (D7), eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-5981-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15237657

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Description: The D7 monoclonal antibody reacts with mouse Sca-1, an 18 kDa member of the Ly-6 family of GPI-linked surface proteins. D7 reacts with both Ly-6E.1 and Ly-6A.2 molecules expressed by mouse hematopoietic stem cells, myeloid population and peripheral T and B cells. Non-activated splenocytes from Ly-6.2 strains including C57BL, SJL, 129, AKR, and others demonstrate higher frequency of D7-positive cells compared to the Ly-6.1 strains such as BALB/c, C3H, NZB and others. Sca-1 is expressed at high levels upon activation regardless of the Ly-6 haplotype. The Ly-6 family is involved in regulation and function of T cell activation. Sca-1 is a major phenotypic marker for mouse hematopoietic progenitor/stem cell subset. Applications Reported: This D7 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and western blotting. Applications Tested: This D7 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. (Fluorochrome-conjugated D7 is recommended for use in flow cytometry.).

Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered. Ly-6A/E molecules are implicated in the regulation of T cell activation.
Specifications

Specifications

Ly-6A/E (Sca-1)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P05533, Q64253
Ly6a, Ly6e
Affinity chromatography
RUO
110454, 17069
4° C
Liquid
Flow Cytometry, Immunoprecipitation, Western Blot
D7
Unconjugated
Ly6e
lymphocyte antigen 6 complex, locus A/E
Rat
50 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.