missing translation for 'onlineSavingsMsg'
Learn More

Ly-49I, PE, clone: YLI-90, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5895-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15278889

  • £68.96 / 50µg


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The YLI-90 monoclonal antibody reacts with the B6 mouse Ly-49I receptor. Ly49I, a member of the Ly49 family with inhibitory function, is a homodimer type II transmembrane protein and contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM). YLI-90 binds to subpopulations of NK, NKT and T cell lineages in the C57Bl/6 strain. This antibody does not bind to cells from BALB/c, SJL or AKR/J mice.Applications Reported: YLI-90 has been reported for use in flow cytometric analysis.Applications Tested: The YLI-90 antibody has been tested by flow cytometric analysis of C57Bl/6 and BALB/c mouse splenocytes and can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ly-49I
YLI-90
PE
Conjugated
Murine
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Ly49I
IgG1
RUO
Primary
Murine
Videos