missing translation for 'onlineSavingsMsg'
Learn More

Ly-49H, Biotin, clone: 3D10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5886-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209899

  • £49.99 / 50µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The monoclonal antibody 3D10 recognizes Ly-49H, a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly-49H, like Ly-49D, lacks the characteristic ITIM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly-49H is expressed in a subset of NK cells but not NKT (NK1.1CD3+) cells. Natural killer cells can express several Ly-49 proteins. It has been shown that there is preferential staining of Ly-49H with Ly-49D (cat 12-5783). Expression has been confirmed on C57BL/6 and NWNA but not Balb/c or DBA/2 mice. Other mice strains have not been tested. No crossreactivity to Ly-49A, C, D or G2 has been observed. Addition of monoclonal antibody 3D10 to cell cultures induces lysis of target cells.Applications Reported: This 3D10 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 3D10 antibody has been tested by flow cytometric analysis of C57Bl/6 mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ly-49H
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q60682
Klra8
IgG1, kappa
50μg
Store at 2-8°C. Do not freeze.
Primary
16639
Flow Cytometry
3D10
Biotin
Klra8
Conjugated
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Videos

For Research Use Only.