Learn More
Ly-49D Rat anti-Mouse, PerCP-eFluor 710, Clone: eBio4E5 (4E5), eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 46-5782-82
Code : NEW
Additional Details : Weight : 0.09500kg
 View more versions of this product
Product Code. 15539276
  • £195.53 / 100µg
Estimated Shipment date
from Supplier 02-07-2018
Add to basket
Description
Description
The monoclonal antibody eBio4E5 (4E5) reacts with Ly-49D, an activating member of the Ly-49 family of NK cell receptors. This family of receptors form homodimers that recognize MHC Class I molecules. The activating Ly-49 receptors (Ly-49D and Ly-49H) do not contain immune receptor tyrosine-based inhibitory motifs (ITIM) in their cytoplasmic domains, distinguishing them from the inhibitory Ly-49 family members. Instead, activating Ly-49 molecules have been shown to associate with DAP12 which contains an immunoreceptor tyrosine-based activation motif (ITAM) that is required for positive signaling. Expression of different Ly-49 family members is not linked, therefore, several Ly-49 proteins can be expressed on the same cells. Expression of Ly-49D on subsets of NK cells is strain-dependent and is found in C57BL/6, CB6F1 and SJL mice, while BALB/c, DBA/2, AKR and CBA/J mice are negative for Ly-49D. In 129/J mice, eBio4E5 also recognizes Ly-49O, Ly-49V and Ly-49R. eBio4E5 is reported to have functional (activating) activity.
Specifications
Specifications
Ly-49D
eBio4E5 (4E5)
PerCP-eFluor 710
Conjugated
IgG2a, kappa
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.