missing translation for 'onlineSavingsMsg'
Learn More

Ly-49C/I/F/H, FITC, clone: 14B11, eBioscience™

Syrian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5991-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15218329

  • £176.50 / 500µg


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Description

Description

Description: The 14B11 monoclonal antibody reacts with a subset of mouse Ly-49 receptors and recognizes a majority of mouse NK cells. This antibody reacts with 3 putative inhibitory members of the Ly-49 family, F, I, and C and one putative activating receptor, Ly-49H. 14B11 does not react with Ly-49A, B, D, or G. It is reported that 14B11 enhances the redirected lysis of FcR+ target cells by IL-2-activated NK cells.Applications Reported: The 14B11 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 14B11 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ly-49C/I/F/H
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q60653, Q64329, Q60682
Killer cell lectin-like receptor 3, 5E6, Lymphocyte antigen 49c, Nk2.1, T-cell surface glycoprotein Ly-49C, Klra3, Ly-49c, Ly49C, Killer cell lectin-like receptor 6, Lymphocyte antigen 49f, Ly-49f, T-cell surface glycoprotein Ly-49F, Klra6, Ly-49f, Ly49-f, Ly49F
Golden Syrian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
14B11
FITC
Klra8
Conjugated
Klra3, Klra6, Klra8
IgG
500μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16634, 16637, 16639
Videos

For Research Use Only.