missing translation for 'onlineSavingsMsg'
Learn More

LY 456236 hydrochloride, Tocris Bioscience™

Selective mGlu1 antagonist

Brand:  Tocris (Bio-Techne) 239010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461124

  • £128.25 / 10mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Selective mGlu1 receptor antagonist (IC50 values are 143 nM and > 10 μM for mGlu1 and mGlu5 receptors respectively). Reduces hyperalgesic behavior induced by formalin in both mouse and rat with ED50 values of 28 and 16.3 mg/kg respectively.
Specifications

Chemical Identifiers

338736-46-2
317.773
ly 456236 hydrochloride, mpmq hydrochloride, 6-methoxy-n-4-methoxyphenyl quinazolin-4-amine hydrochloride, 4-methoxyphenyl-6-methoxyquinazolin-4-yl amine hydrochloride, 4-methoxy-phenyl-6-methoxy-quinazolin-4-yl-amine hydrochloride, 6-methoxy-n-4-methoxyphenyl-4-quinazolinamine hydrochloride
6-methoxy-N-(4-methoxyphenyl)quinazolin-4-amine;hydrochloride
C16H16ClN3O2
AVKFOWUSTVWZQN-UHFFFAOYSA-N
9926998
COC1=CC=C(C=C1)NC2=NC=NC3=C2C=C(C=C3)OC.Cl

Specifications

LY 456236 hydrochloride
338736-46-2
ly 456236 hydrochloride, mpmq hydrochloride, 6-methoxy-n-4-methoxyphenyl quinazolin-4-amine hydrochloride, 4-methoxyphenyl-6-methoxyquinazolin-4-yl amine hydrochloride, 4-methoxy-phenyl-6-methoxy-quinazolin-4-yl-amine hydrochloride, 6-methoxy-n-4-methoxyphenyl-4-quinazolinamine hydrochloride
COC1=CC=C(C=C1)NC2=NC=NC3=C2C=C(C=C3)OC.Cl
317.773
317.77
10mg
C16H16ClN3O2
AVKFOWUSTVWZQN-UHFFFAOYSA-N
6-methoxy-N-(4-methoxyphenyl)quinazolin-4-amine;hydrochloride
9926998
>99%
Videos