Learn More
LY 367385, Tocris Bioscience™
Selective mGlu1a antagonist
Brand:  Tocris (Bio-Techne) 1237/50
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488933
  • £600.29 / 50mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
A selective mGlu1a receptor antagonist, with an IC50 value of 8.8 μM for blockade of quisqualate-induced phosphoinositide hydrolysis vs. > 100 μM for mGlu5a, and negligible action on group II and III receptors. Also available as part of the Group I mGlu Receptor Tocriset™ .
Specifications
Chemical Identifiers
198419-91-9
209.201
(S)-(+)-α-Amino-4-carboxy-2-methylbenzeneacetic acid
4-[(S)-amino(carboxy)methyl]-3-methylbenzoic acid
C10H11NO4
SGIKDIUCJAUSRD-QMMMGPOBSA-N
5311261
CC1=C(C=CC(=C1)C(=O)O)C(C(=O)O)N
Specifications
LY 367385
50mg
C10H11NO4
SGIKDIUCJAUSRD-QMMMGPOBSA-N
4-[(S)-amino(carboxy)methyl]-3-methylbenzoic acid
5311261
198419-91-9
>99%
(S)-(+)-α-Amino-4-carboxy-2-methylbenzeneacetic acid
CC1=C(C=CC(=C1)C(=O)O)C(C(=O)O)N
209.201
209.2
Videos