missing translation for 'onlineSavingsMsg'
Learn More

LY 344864 hydrochloride, Tocris Bioscience™

Potent, selective 5-HT1F agonist

Brand:  Tocris (Bio-Techne) 245110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461234

  • £178.20 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Potent, selective 5-HT1F receptor agonist (EC50 = 3 nM). Displays > 80-fold selectivity over other 5-HT receptors (Ki values are 0.006, 0.53, 0.55, 0.56, 1.42, 1.70, 3.50, 3.94 and 4.85 μM for 5-HT1F, 5-HT1A, 5-HT1B, 5-HT1D, 5-HT1E, 5-HT2B, 5-HT2C, 5-HT2A and 5-HT7 receptors respectively). Inhibits neurogenic dural inflammation in vivo following i.v. and oral administration.
Specifications

Chemical Identifiers

1217756-94-9
387.883
r-n-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluorobenzamide hydrochloride, ly-344864 hydrochloride, ly 344864 hydrochloride, n-6r-6-dimethylamino-6,7,8,9-tetrahydro-5h-carbazol-3-yl-4-fluorobenzamide hydrochloride, n-3r-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluoro-benzamide hcl, n-3r-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluorobenzamidehydrochloride
N-[(6R)-6-(dimethylamino)-6,7,8,9-tetrahydro-5H-carbazol-3-yl]-4-fluorobenzamide;hydrochloride
C21H23ClFN3O
OKUHLSYESBLBCP-PKLMIRHRSA-N
56972176
CN(C)C1CCC2=C(C1)C3=C(N2)C=CC(=C3)NC(=O)C4=CC=C(C=C4)F.Cl

Specifications

LY 344864 hydrochloride
1217756-94-9
r-n-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluorobenzamide hydrochloride, ly-344864 hydrochloride, ly 344864 hydrochloride, n-6r-6-dimethylamino-6,7,8,9-tetrahydro-5h-carbazol-3-yl-4-fluorobenzamide hydrochloride, n-3r-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluoro-benzamide hcl, n-3r-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluorobenzamidehydrochloride
CN(C)C1CCC2=C(C1)C3=C(N2)C=CC(=C3)NC(=O)C4=CC=C(C=C4)F.Cl
387.883
387.88
10mg
C21H23ClFN3O
OKUHLSYESBLBCP-PKLMIRHRSA-N
N-[(6R)-6-(dimethylamino)-6,7,8,9-tetrahydro-5H-carbazol-3-yl]-4-fluorobenzamide;hydrochloride
56972176
>99%
Videos