Learn More
LY 341495 disodium salt, Tocris Bioscience™
Potent and selective group II mGlu antagonist; disodium salt of LY 341495 (Cat. No. 1209)
Brand:  RD Systems (Bio-Techne) 4062/10
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 10mg
 View more versions of this product
Product Code. 13464064
  • £257.25 / 10mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Highly potent and selective group II metabotropic glutamate receptor antagonist. Disodium salt of LY 341495 (Cat. No. 1209).
Specifications
Chemical Identifiers
C20H17NNa2O5
(2S)-2-Amino-2-[(1S, 2S)-2-carboxycycloprop-1-yl]-3-(xanth-9-yl) propanoic acid disodium salt
Specifications
10mg
C20H17NNa2O5
397.33
LY 341495 disodium salt
(2S)-2-Amino-2-[(1S, 2S)-2-carboxycycloprop-1-yl]-3-(xanth-9-yl) propanoic acid disodium salt
>97%