Learn More
LY 294002 hydrochloride, Tocris Bioscience™
Prototypical PI 3-kinase inhibitor; also inhibits other kinases
Brand:  Tocris (Bio-Techne) 113025
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 25mg
 View more versions of this product
Product Code. 13488623
  • £448.52 / 25mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Prototypical PI 3-kinase inhibitor (IC50 values are 0.31, 0.73, 1.06 and 6.60 μM for PI 3-Kβ, PI 3-Kα, PI 3-Kδ and PI 3-Kγ respectively). Also inhibits other kinases including, CK2, mTOR, PLK1, PIM1 and PIM3. Inhibits proliferation and induces apoptosis in human colon cancer cells in vitro and in vivo. Suppresses proliferation of mESCs. Inhibits autophagic sequestration in rat hepatocytes. Negative control LY 303511 (Cat. No. 2418) also available.
Specifications
Chemical Identifiers
934389-88-5
343.807
2-(4-Morpholinyl)-8-phenyl-4H-1-benzopyran-4-one hydrochloride
C19H18ClNO3
OQZQSRICUOWBLW-UHFFFAOYSA-N
11957589
Specifications
LY 294002 hydrochloride
>99%
2-(4-Morpholinyl)-8-phenyl-4H-1-benzopyran-4-one hydrochloride
C1COCCN1C2=CC(=O)C3=C(O2)C(=CC=C3)C4=CC=CC=C4.Cl
343.807
343.81
934389-88-5
C19H18ClNO3
OQZQSRICUOWBLW-UHFFFAOYSA-N
2-morpholin-4-yl-8-phenylchromen-4-one;hydrochloride
11957589
25mg
Videos