missing translation for 'onlineSavingsMsg'
Learn More

LY 255283, Tocris Bioscience™

Selective, competitive BLT2 receptor antagonist

Brand:  Tocris (Bio-Techne) 220810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13440804

  • £189.63 / 10mg

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective, competitive antagonist of BLT2 receptors (IC50 values are ∽ 1 and > 10 μM at human recombinant BLT2 and BLT1 receptors respectively). Inhibits LTB4-induced contraction of lung parenchyma (pA2 = 7.2), and reduces LTB4-mediated airway obstruction in guinea pigs following i.v. and oral administration. .
Specifications

Chemical Identifiers

117690-79-6
360.458
unii-h037w1i5al, chembl15766, 1-5-ethyl-2-hydroxy-4-6-methyl-6-1h-tetrazol-5-yl heptyloxy phenyl ethanone, 1-5-ethyl-2-hydroxy-4-6-methyl-6-1h-tetrazol-5-yl heptyl oxy phenyl ethanone, ethanone, 1-5-ethyl-2-hydroxy-4-6-methyl-6-1h-tetrazol-5-yl heptyl oxy phenyl, 1-5-ethyl-2-hydroxy-4-6-methyl-6-1h-1,2,3,4-tetrazol-5-yl heptyl oxy phenyl ethanone, ly hplc , powder, 1-5-ethyl-2-hydroxy-4-6-methyl-6-2h-tetrazol-5-yl heptoxy phenyl ethanone
1-[5-ethyl-2-hydroxy-4-[6-methyl-6-(2H-tetrazol-5-yl)heptoxy]phenyl]ethanone
C19H28N4O3
WCGXJPFHTHQNJL-UHFFFAOYSA-N
122023
CCC1=C(C=C(C(=C1)C(=O)C)O)OCCCCCC(C)(C)C2=NNN=N2

Specifications

10mg
117690-79-6
unii-h037w1i5al, chembl15766, 1-5-ethyl-2-hydroxy-4-6-methyl-6-1h-tetrazol-5-yl heptyloxy phenyl ethanone, 1-5-ethyl-2-hydroxy-4-6-methyl-6-1h-tetrazol-5-yl heptyl oxy phenyl ethanone, ethanone, 1-5-ethyl-2-hydroxy-4-6-methyl-6-1h-tetrazol-5-yl heptyl oxy phenyl, 1-5-ethyl-2-hydroxy-4-6-methyl-6-1h-1,2,3,4-tetrazol-5-yl heptyl oxy phenyl ethanone, ly hplc , powder, 1-5-ethyl-2-hydroxy-4-6-methyl-6-2h-tetrazol-5-yl heptoxy phenyl ethanone
CCC1=C(C=C(C(=C1)C(=O)C)O)OCCCCCC(C)(C)C2=NNN=N2
360.458
360.46
LY 255283
C19H28N4O3
WCGXJPFHTHQNJL-UHFFFAOYSA-N
1-[5-ethyl-2-hydroxy-4-[6-methyl-6-(2H-tetrazol-5-yl)heptoxy]phenyl]ethanone
122023
>98%
Videos
Documents