Learn More
Tocris Bioscience™ LY 233053
Brand:  Tocris Bioscience™ 152110
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13449603
  • £189.63 / 10mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Competitive NMDA receptor antagonist (IC50 = 7nM) that displays no affinity for AMPA or kainate receptors at a concentration of 10μM. Inhibits NMDA-induced neuronal degeneration and protects from NMDA-induced convulsions in neonatal rats.
Specifications
Specifications
(2R,4S)-4-(1H-Tetrazol-5-ylmethyl)-2-piperidinecarboxylic acid
>99%
Observed MW: 211.22
Soluble to 100mM in DMSO
125546-04-5
C8H13N5O2