missing translation for 'onlineSavingsMsg'
Learn More

LY 215840, Tocris Bioscience™

5-HT2/5-HT7 antagonist

Brand:  Tocris (Bio-Techne) 152310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13469603

  • £158.76 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

5-HT2/5-HT7 receptor antagonist. Reduces stimulatory effect of serotonin on aldosterone secretion; also reduces 5-HT-induced cAMP formation (pKB = 8.26).
Specifications

Chemical Identifiers

137328-52-0
395.547
[8β(1S, 2R)]-N-(2-Hydroxycyclopentyl)-6-methyl-1-(1-methylethyl)-ergoline-8-carboxamide
(6aR,9R,10aS)-N-[(1S,2R)-2-hydroxycyclopentyl]-7-methyl-4-propan-2-yl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-9-carboxamide
C24H33N3O2
IMSDOBUYDTVEHN-ZLJPILQHSA-N
122173083
CC(C)N1C=C2CC3C(CC(CN3C)C(=O)NC4CCCC4O)C5=C2C1=CC=C5

Specifications

10mg
137328-52-0
[8β(1S, 2R)]-N-(2-Hydroxycyclopentyl)-6-methyl-1-(1-methylethyl)-ergoline-8-carboxamide
CC(C)N1C=C2CC3C(CC(CN3C)C(=O)NC4CCCC4O)C5=C2C1=CC=C5
395.547
395.54
LY 215840
C24H33N3O2
IMSDOBUYDTVEHN-ZLJPILQHSA-N
(6aR,9R,10aS)-N-[(1S,2R)-2-hydroxycyclopentyl]-7-methyl-4-propan-2-yl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-9-carboxamide
122173083
>98%
Videos
Documents