missing translation for 'onlineSavingsMsg'
Learn More

LY 2087101, Tocris Bioscience™

Potentiator of α7 nAChRs

Brand:  Tocris (Bio-Techne) 414150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444234

  • £613.09 / 50mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Allosteric potentiator of α7, α4β2 and α4β4 nAChRs; displays selectivity against α3β4 nAChRs. Thought to potentiate agonist-evoked α7 responses by binding within the nAChR transmembrane region.
Specifications

Chemical Identifiers

913186-74-0
318.384
d04mig, 2-4-fluorophenylamino-4-methyl-thiazol-5-yl-thiophen-3-yl-methanone, 2-4-fluorophenyl amino-4-methyl-5-thiazolyl-3-thienylmethanone, methanone, 2-4-fluorophenyl amino-4-methyl-5-thiazolyl-3-thienyl, n-4-fluorophenyl-4-methyl-5-thiophen-3-yl carbonyl-1,3-thiazol-2-amine
[2-(4-fluoroanilino)-4-methyl-1,3-thiazol-5-yl]-thiophen-3-ylmethanone
C15H11FN2OS2
PEAMDZVDNYENPN-UHFFFAOYSA-N
11964458
CC1=C(SC(=N1)NC2=CC=C(C=C2)F)C(=O)C3=CSC=C3

Specifications

LY 2087101
913186-74-0
d04mig, 2-4-fluorophenylamino-4-methyl-thiazol-5-yl-thiophen-3-yl-methanone, 2-4-fluorophenyl amino-4-methyl-5-thiazolyl-3-thienylmethanone, methanone, 2-4-fluorophenyl amino-4-methyl-5-thiazolyl-3-thienyl, n-4-fluorophenyl-4-methyl-5-thiophen-3-yl carbonyl-1,3-thiazol-2-amine
CC1=C(SC(=N1)NC2=CC=C(C=C2)F)C(=O)C3=CSC=C3
318.384
318.39
50mg
C15H11FN2OS2
PEAMDZVDNYENPN-UHFFFAOYSA-N
[2-(4-fluoroanilino)-4-methyl-1,3-thiazol-5-yl]-thiophen-3-ylmethanone
11964458
>98%
Videos