missing translation for 'onlineSavingsMsg'
Learn More

Ly-108, clone: eBio13G3-19D (13G3-19D), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-1508-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15247247

  • £157.52 / 100µg


Estimated Shipment
06-07-2018
Add to basket

Description

Description

Description: The eBio13G3-19D monoclonal antibody reacts with mouse Ly108, the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is also known as NTB-A (NK cell, T cell, B cell antigen), and is expressed on NK, T cells, and B cells. Engagement of the Ly108 receptor enhances NK cell activity and expression of IFN-gamma, and contributes to the inability of NK cells to kill Epstein Barr virus-infected B cells in X-linked lymphoproliferative disease. CD4+ T cells from Ly108 knock-out mice exhibit diminished ability to produce IL-4 and defective neutrophil functions.Staining has not revealed any strain differences between C57BL/6 and BALB/c mice. Other strains have not been tested.Applications Reported: This eBio13G3-19D (13G3-19D) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB).Applications Tested: This eBio13G3-19D (13G3-19D) antibody has been tested by flow cytometric analysis of mouse splenocytes and thymocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ly-108
Monoclonal
0.5mg/mL
TBS with 0.09% sodium azide; pH 8.0
Q9ET39
SLAM family member 6, Activating NK receptor, NK-T-B-antigen, NTB-A, CD352, SLAMF6, KALI, UNQ6123/PRO20080
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryþImmunoprecipitationþWestern Blot
eBio13G3-19D (13G3-19D)
Unconjugated
Slamf6
Purified
Slamf6
IgG2a, kappa
100μg
Store at 2-8°C.
Primary
30925
Videos

For Research Use Only.