missing translation for 'onlineSavingsMsg'
Learn More

Luzindole, Tocris Bioscience™

Competitive melatonin MT1/MT2 antagonist

Brand:  Tocris (Bio-Techne) 087710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13468093

  • £170.52 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Melatonin antagonist.
Specifications

Chemical Identifiers

117946-91-5
292.382
luzindole, n-acetyl-2-benzyltryptamine, 2-benzyl-n-acetyltryptamine, n-2-2-benzyl-1h-indol-3-yl ethyl acetamide, n-2-2-phenylmethyl-1h-indol-3-yl ethyl acetamide, tocris-0877, n-acetyl-2-benzyl-tryptamine, acetamide, n-2-2-phenylmethyl-1h-indol-3-yl ethyl, n-2-2-benzylindol-3-yl ethyl acetamide
N-[2-(2-benzyl-1H-indol-3-yl)ethyl]acetamide
C19H20N2O
WVVXBPKOIZGVNS-UHFFFAOYSA-N
122162
CC(=O)NCCC1=C(NC2=CC=CC=C21)CC3=CC=CC=C3

Specifications

10mg
117946-91-5
luzindole, n-acetyl-2-benzyltryptamine, 2-benzyl-n-acetyltryptamine, n-2-2-benzyl-1h-indol-3-yl ethyl acetamide, n-2-2-phenylmethyl-1h-indol-3-yl ethyl acetamide, tocris-0877, n-acetyl-2-benzyl-tryptamine, acetamide, n-2-2-phenylmethyl-1h-indol-3-yl ethyl, n-2-2-benzylindol-3-yl ethyl acetamide
CC(=O)NCCC1=C(NC2=CC=CC=C21)CC3=CC=CC=C3
292.382
292.38
Luzindole
C19H20N2O
WVVXBPKOIZGVNS-UHFFFAOYSA-N
N-[2-(2-benzyl-1H-indol-3-yl)ethyl]acetamide
122162
>99%
Videos
Documents