missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lutetium rod, 6.35mm (0.25 in.) dia., 99.9% (metals basis excluding Ta), Ta typically 2%

Brand:  Alfa Aesar™ 043961.HF

Code : NEW

Additional Details : CAS Number : 7439-94-3 Weight : 0.01000kg

Quantity 10mm

 View more versions of this product

Product Code. 11395008

  • £317.15 / 10mm


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Specifications

Chemical Identifiers

Lutetium rod
7439-94-3
≈6.35mm (0.25 in.) dia.
MFCD00011098

Specifications

Lutetium rod
≈6.35mm (0.25 in.) dia.
Odorless
Cast Rod
(metals basis excluding Ta), Ta typically 2%
10mm
7439-94-3
Mylar under Argon
99.9%
MFCD00011098
Videos
SDS