missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lutetium, plasma standard solution, Specpure™, Lu 10,000μg/mL

Brand:  Alfa Aesar™ 035757.AD

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 50mL

 View more versions of this product

Product Code. 11308647

  • £179.70 / 50mL


Estimated Shipment date
from Supplier 18-07-2018
Add to basket

Specifications

Chemical Identifiers

Lutetium plasma standard solution
Specpure™
UN3264
Lu 10000μg/mL
MFCD00011098

Specifications

Lutetium plasma standard solution
Specpure™
Lu2O3 in 5% HNO3
UN3264
Lu 10000μg/mL
Liquid
MFCD00011098
50mL
Videos
SDS