missing translation for 'onlineSavingsMsg'
Learn More

Lutetium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7048998
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7439-94-3
Lu
174.97
MFCD00011098
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
23929
CHEBI:33382
lutetium
[Lu]
This item is not returnable. View return policy
7439-94-3
Lu
174.97
MFCD00011098
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
23929
CHEBI:33382
lutetium
[Lu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Silver
UN Number UN3178
Quantity 1 g
Percent Purity 99.9%
Odor Odorless
Form Pieces
Assay (REO)
Chemical Name or Material Lutetium pieces, Distilled dendritic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'