missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lutetium(III) trifluoromethanesulfonate, 98%

Brand:  Alfa Aesar™ H60170.MD

Code : NEW

Additional Details : CAS Number : 126857-69-0 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15416648

  • £26.39 / 250mg


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Description

Description

Applications
It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Hygroscopic. Keep away from strong oxidizing agents. Store at room temperature.
Specifications

Chemical Identifiers

Lutetium(III) trifluoromethanesulfonate
C3F9LuO9S3
622.17
126857-69-0
(CF3SO3)3Lu
MFCD00192254

Specifications

Lutetium(III) trifluoromethanesulfonate
98%
(CF3SO3)3Lu
MFCD00192254
126857-69-0
C3F9LuO9S3
622.17
250mg
Videos
SDS