missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lutetium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Lutetium Sulfate also called Lutetium Sulphate, is applied in making phosphors. Stable Lutetium can be used as catalysts in petroleum cracking in refineries and can also be used in alkylation, hydrogenation, and polymerization applications. It can also be used as an ideal host for X-ray phosphors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline |
Quantity | 1 g |
Merck Index | 14,5615 |
Solubility Information | Soluble in water,moderately soluble in strong mineral acids. |
Formula Weight | 782.24 (683.12 Anhydrous) |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Assay | (REO) |
Chemical Name or Material | Lutetium(III) sulfate octahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'