missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) oxide, REacton™, 99.995% (metals basis), REM 50ppm, Thermo Scientific Chemicals

Catalog Number 11321438
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
12032-20-1
Lu2O3
397.93
MFCD00011100
UGBIHFMRUDAMBY-UHFFFAOYSA-N
lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
dilutetium(3+) trioxidandiide
[O--].[O--].[O--].[Lu+3].[Lu+3]
This item is not returnable. View return policy
12032-20-1
Lu2O3
397.93
MFCD00011100
UGBIHFMRUDAMBY-UHFFFAOYSA-N
lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
dilutetium(3+) trioxidandiide
[O--].[O--].[O--].[Lu+3].[Lu+3]

The product of Lutetium(III) oxide and palladium(II) oxide has a potential use in superconducting materials. It is an important raw material for laser crystals. It has specialized uses in ceramics, glass, phosphors, and lasers. Also used as a catalyst in cracking, alkylation, hydrogenation, and polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2487°C
Physical Form Powder
Quantity 5 g
Merck Index 14,5615
Solubility Information Insoluble in water.
Formula Weight 397.94
Percent Purity 99.995%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 9.420 g/mL
Assay (metals basis)
Chemical Name or Material Lutetium(III) oxide, REM 50ppm
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'