missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number 11316328
Click to view available options
Quantity:
2 g
10 g
100641-16-5
LuN3O9
360.98
MFCD00150421
APRNQTOXCXOSHO-UHFFFAOYSA-N
lutetium iii nitrate hydrate, lutetium nitrate hydrate, acmc-1buji, lu.3no3.h2o, lutetium nitrate-water 1/3/1, lutetium 3+ hydrate trinitrate
16212852
lutetium(3+);trinitrate;hydrate
[Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
100641-16-5
LuN3O9
360.98
MFCD00150421
APRNQTOXCXOSHO-UHFFFAOYSA-N
lutetium iii nitrate hydrate, lutetium nitrate hydrate, acmc-1buji, lu.3no3.h2o, lutetium nitrate-water 1/3/1, lutetium 3+ hydrate trinitrate
16212852
lutetium(3+);trinitrate;hydrate
[Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline Aggregates
Quantity 10 g
Linear Formula Lu(NO3)3·xH2O
UN Number UN1477
Formula Weight 360.98 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'