missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific™

Catalog Number p-7050190
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13813-45-1
MFCD00049970
This item is not returnable. View return policy
13813-45-1
MFCD00049970

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 1050°C
Physical Form Flakes or Powder
Quantity 1 g
Solubility Information Soluble in chloroform,carbon tetrachloride and carbon disulfide.
Formula Weight 555.68
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Density 5.60 g/mL
Assay (REO)
Chemical Name or Material Lutetium(III) iodide, Ultra dry
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'