missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059331
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13760-81-1
LuF3
MFCD00016191
This item is not returnable. View return policy
13760-81-1
LuF3
MFCD00016191

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1182°C
Physical Form Powder
Quantity 1 g
UN Number UN3288
Formula Weight 231.97
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 8.3 g/mL
Assay (metals basis)
Chemical Name or Material Lutetium (III) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'