missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific™

Catalog Number p-3212830
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10099-66-8
Cl3Lu
281.33
MFCD00011099
This item is not returnable. View return policy
10099-66-8
Cl3Lu
281.33
MFCD00011099
For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 905°C
Color White to Gray
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9%
Linear Formula LuCl3
Solubility Information Solubility in water: soluble
Formula Weight 281.33
Percent Purity 99.90%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lutetium(III) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Lutetium(III) chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'