missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lutetium(III) carbonate hydrate, REacton™, 99.9% (REO)

Brand:  Alfa Aesar™ 014569.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11332518

  • £90.74 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Lutetium(III) carbonate hydrate
Reacton™
529.97 (Anhydrous)
MFCD00799559

Specifications

Lutetium(III) carbonate hydrate
Crystalline
(REO)
MFCD00799559
1g
Reacton™
99.9%
529.97 (Anhydrous)
Hygroscopic
Videos
SDS