missing translation for 'onlineSavingsMsg'
Learn More

Lutetium, AAS standard solution, Specpure™ Lu 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4408565
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
12032-20-1, 7697-37-2, 7732-18-5
Lu2O3
397.93
MFCD00011100
UGBIHFMRUDAMBY-UHFFFAOYSA-N
dilutetium(3+) trioxidandiide
[O--].[O--].[O--].[Lu+3].[Lu+3]
This item is not returnable. View return policy
12032-20-1, 7697-37-2, 7732-18-5
Lu2O3
397.93
MFCD00011100
UGBIHFMRUDAMBY-UHFFFAOYSA-N
dilutetium(3+) trioxidandiide
[O--].[O--].[O--].[Lu+3].[Lu+3]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mL
Grade AAS Standard
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Lutetium(III) oxid: 0.11%; Water: 94.89%
Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Chemical Name or Material Lutetium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'