Learn More
Thermo Scientific™ Luminaris Color Probe qPCR Master Mix
Optimize qPCR with probe detection using standard cycling protocols with these ready-to-use master mixes with premixed UDG, available with or without blue color.
Brand: Thermo Scientific™ K0354
2196.77 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 88
Additional Details : Weight : 0.22000kg
Description
Luminaris Color Probe and Luminaris Probe qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR. The master mixes include Thermo Scientific Hot Start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimized PCR buffer. Only the template, primers and probe need to be added.
The Hot Start Taq DNA polymerase, in combination with an optimized buffer, ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control. The Luminaris Color Probe qPCR Master Mixes are supplemented with an inert blue dye and a separate Yellow Sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.
The use of Luminaris Color Probe and Luminaris Probe qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color Probe and Luminaris Probe qPCR Master Mixes with ROX at different concentrations as passive reference dye are available for most qPCR platforms.
Thermo Scientific Luminaris Color Probe qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR. The master mixes include Thermo Scientific Hot Start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimized PCR buffer. Only the template, primers and probe need to be added.The Hot Start Taq DNA polymerase, in combination with an optimized buffer, ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control. The Luminaris Color Probe qPCR Master Mixes are supplemented with an inert blue dye and a separate Yellow Sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.
The use of Luminaris Color Probe qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color Probe qPCR Master Mixes with ROX at different concentrations as passive reference dye are available for most qPCR platforms.
Specificity no primer dimers
Sensitivitity detects low copy number targets
Wide linear range accurate quantification across 9 orders of magnitude
Reproducibility and convenience ready-to-use 2X master mix
- Blue Master Mix and Yellow Sample Buffer for easy pipetting
- UDG in the master mix to prevent carry-over contamination
- Excellent qPCR performance
- Versions with premixed ROX for different real-time platforms
- Standard cycling protocols
Includes:
- Luminaris Color Probe qPCR Master Mixes include Hot Start Taq Polymerase, UDG and dNTPs (also dUTP) in an optimized PCR buffer with a blue dye
- 40X Yellow Sample Buffer with a yellow dye
- Nuclease-free water
Recommended for:
Gene expression analysis; siRNA validation; Genotyping; Pathogen detection
Specifications
Contains: • 4 x 12.5 mL Luminaris Color Probe qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG and dNTPs (with dUTP) in an optimized PCR buffer with a blue dye; sufficient for 5000 x 20 μL reactions • 1 x 13 mL 40X Yellow Sample Buffer • 2 x 30 mL Nuclease-Free Water Store at -20°C. |
|
Primer-probe | |
High | |
Taq DNA Polymerase | |
BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon | |
qPCR Master Mix | |
5000 Reactions of 20 μL | |
2X (Luminaris Color Probe qPCR Master Mix), 40X (Yellow Sample Buffer) | |
DNA, dsDNA |
Color Probe | |
Tube | |
qPCR | |
Standard | |
Luminaris | |
5000 x 20 μL Reactions | |
Gene Expression | |
5,000 Reactions | |
4 x 12.5 mL |
For Research Use Only. Not for use in diagnostic procedures.