missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Luminaris Color HiGreen qPCR Master Mix, fluorescein
Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.
£218.00 - £3170.00
Specifications
Detection Method | SYBR |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
13283179
|
Thermo Scientific™
K0381 |
250 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13203189
|
Thermo Scientific™
K0383 |
1250 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13213189
|
Thermo Scientific™
K0384 |
5000 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13293179
|
Thermo Scientific™
K0382 |
500 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ Luminaris Color HiGreen Master Mixes are ready-to-use qPCR master mixes with color and premixed UDG optimized for SYBR Green chemistry using standard cycling protocols.
- Blue master mix and Yellow Sample Buffer for easy pipetting
- UDG in the master mix to prevent carry-over contamination
- Equal performance of blue and colorless master mixes
- Versions with premixed ROX or fluorescein for different real-time platforms
- Standard cycling protocols
- Excellent qPCR performance
- Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity: single copy detection
- Wide linear range: accurate quantification across 9 orders of magnitude
- Reproducibility and convenience: ready-to-use 2X master mix
Specifications
SYBR | |
qPCR | |
Standard | |
Luminaris | |
Gene Expression | |
2 X |
High | |
Taq DNA Polymerase | |
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 | |
qPCR Master Mix | |
2X | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.