Learn More
LUC7L3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553816
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LAEVERRIRR GHARLALSQN QQSSGAAGPT GKNEEKIQVL TDKIDVLLQQ IEELGSEGKV EEAQGMMKLV EQLKEERELL RSTTSTIESF A Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
LUC7L3 Polyclonal antibody specifically detects LUC7L3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
LUC7L3 | |
Polyclonal | |
Unconjugated | |
LUC7L3 | |
3300001P08Rik; cAMP regulatory element-associated protein 1; cisplatin resistance associated overexpressed protein; cisplatin resistance-associated overexpresse protein-like protein; cisplatin resistance-associated overexpressed protein; cisplatin resistance-associated-overexpressed protein; CRA; CREAP1; CREAP-1; CRE-associated protein 1; Crop; FLJ11063; hLuc7A; LUC7 like 3 pre-mRNA splicing factor; Luc7A; LUC7L3; LUC7-like 3; LUC7-like 3 (S. cerevisiae); LUC7-like 3 pre-mRNA splicing factor; luc7-like protein 3; O48; OA48-18; okadaic acid-inducible phosphoprotein OA48-18; RGD1307981; zgc:136380; zgc:158165 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
360602, 51747, 67684 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95232, Q5SUF2 | |
LUC7L3 | |
Recombinant protein corresponding to Human LUC7L3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only