Learn More
Tocris Bioscience™ LU AA33810
Potent NPY Y5 receptor antagonist
Brand:  Tocris Bioscience™ 356210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443204
  • £139.12 / 10mg
Estimated Shipment
14-09-2020
Description
Description
Potent neuropeptide Y (NPY) Y5 receptor antagonist (Ki = 1.5 nM in vitro). Displays ≥ 3300-fold affinity for Y5 over Y1, Y2 and Y4 receptors. Also binds human 5-HT2B and 5-HT1A receptors (Ki values are 247 and 478 nM respectively). Exerts anxiolytic- and antidepressant-like effects in rat models of stress sensitivity.
Specifications
Specifications
LU AA33810
>98
10mg
LU AA33810
423.62