missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LU AA33810
Potent NPY Y5 receptor antagonist
POA
Specifications
Purity | >98% |
---|---|
Product Type | LU AA33810 |
Molecular Weight (g/mol) | 423.62 |
Inhibitors | LU AA33810 |
Description
Potent neuropeptide Y (NPY) Y5 receptor antagonist (Ki = 1.5 nM in vitro). Displays ≥ 3300-fold affinity for Y5 over Y1, Y2 and Y4 receptors. Also binds human 5-HT2B and 5-HT1A receptors (Ki values are 247 and 478 nM respectively). Exerts anxiolytic- and antidepressant-like effects in rat models of stress sensitivity.Specifications
>98% | |
LU AA33810 | |
423.62 | |
LU AA33810 |
For Research Use Only