missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LSECtin/CLEC4G Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | LSECtin/CLEC4G |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
LSECtin/CLEC4G Polyclonal specifically detects LSECtin/CLEC4G in Human samples. It is validated for Western Blot.Specifications
LSECtin/CLEC4G | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CLEC4G, C-type lectin domain family 4 member G, C-type lectin domain family 4, member G, C-type lectin superfamily 4, member G, DTTR431, liver and lymph node sinusoidal endothelial cell C-type lectin, LP2698, LSECtin, Q6UXB4, UNQ431 | |
CLEC4G | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q6UXB4 | |
339390 | |
Mouse myeloma cell line NS0-derived recombinant human LSECtin/CLEC4G Ser54-Cys293 Accession # Q6UXB4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human LSECtin/CLEC4G in direct ELISAs and Western blots. |
For Research Use Only