Learn More
LRTOMT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559005
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MNKRDYMNTS VQEPPLDYSF RSIHVIQDLV NEEPRTGLRP LKRSKSGKSL TQSLWLNNNV LNDLRDFNQV ASQLLEHPEN LAWIDLSFND LTSIDPV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
LRTOMT Polyclonal antibody specifically detects LRTOMT in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
LRTOMT | |
Polyclonal | |
Unconjugated | |
LRTOMT | |
1700008D07Rik; Catechol O-methyltransferase 2; catechol O-methyltransferase 2 {ECO:0000250; CFAP111; Comt2; DFNB63; leucine rich repeat containing 51; leucine rich transmembrane and 0-methyltransferase domain containing; leucine rich transmembrane and O-methyltransferase domain containing; leucine-rich repeat-containing protein 51; leucine-rich repeat-containing protein 51 {ECO:0000312; leucine-rich repeat-containing protein 51; transmembrane O-methyltransferase; LRRC51; lrrc51 {ECO:0000312; Lrtomt; Lrtomt1; O-methyltransferase domain containing; PP7517; Protein LRTOMT1; protein LRTOMT2; RGD:1561509}; RGD:1565856}; RGD1561509; RGD1565856; Tomt; tomt {ECO:0000312; Transmembrane O-methyltransferase; transmembrane O-methyltransferase {ECO:0000312; Transmembrane O-methyltransferase homolog; UniProtKB:A1Y9I9} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220074 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WZ04 | |
LRTOMT | |
Recombinant protein corresponding to Human LRTOMT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only