missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRK2 Rabbit anti-Human, Tb (Terbium), Monoclonal, Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen A14690
Code : NEW
Additional Details : Weight : 0.00900kg
Specifications
LRRK2 | |
Tb (Terbium) | |
Rabbit | |
RUO | |
120892 | |
Store at -20 °C |
Monoclonal | |
NP_940980.3 | |
25 μg | |
Primary | |
Human | |
Liquid |